Learn More
Miles Scientific SILICA GEL HL 250U 20X20CM 25/
SDP
Catalog No. NC0317002
Encompass_Preferred
Silica Gel HL 250um 20x20cm channeled w/Preadsorbent Zone (25 plates/box)
Catalog No. NC0317002 Supplier Miles Scientific Supplier No. P43911
Silica Gel HL 250um 20x20cm channeled w/Preadsorbent Zone (25 plates/box)