Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silica gel, HPLC grade, spherical, 2.2 micron APS, 80 angstroms, 99.99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Spherical, S.A. 470m |
Material | Silica Gel |
Quantity | 5 g |
Particle Size | 2.2 Micron APS, 80Å |
Percent Purity | ≥99.99% |
Grade | HPLC |
Surface Area | 470m2/g |
Chemical Name or Material | Silica gel, P.V. 0.95cc/g |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H373 May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | GHS P Statement P260-P314-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Get medical advice/attention if you feel unwell. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 231-545-4 |
RTECSNumber | VV7340000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction