Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, Indicating Beads Expose to atmosphere only when put into use., Reagents

Catalog No. 01337075
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-075 3 kg
01-337-074 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-075 Supplier Reagents Holdings Llc Supplier No. C23005503A5
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silica Gel, Indicating Beads Expose to atmosphere only when put into use., Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Solid
Color Blue
Material Silica Gel
Quantity 3 kg
Packaging Poly Bottle
Specific Gravity 2.1
Chemical Name or Material Silica Gel, Indicating Beads Expose to atmosphere only when put into use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.