Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel PS Drying Agent, Solstice

Catalog No. 60047560 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-560 1 kg Poly Bottle
60-047-561 250 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-560 Supplier Solstice Supplier No. 783071KG
Add to Cart
Add to Cart

With indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 1 kg
UN Number NONH for all modes of transport
Packaging Poly Bottle
Formula Weight 60.08g/mol
Chemical Name or Material Silica gel PS drying agent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.