Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel Rubin Drying Bags, Solstice

Catalog No. 60047636 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Metal Can
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-636 1 kg Metal Can
60-047-446 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-636 Supplier Solstice Supplier No. 801481KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With indicator function, 1/3 TE, bag with ∼10 g, crystalline

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silica gel Rubin drying bags
CAS 112926-00-8
Material Silica Gel
Quantity 1 kg
Molecular Formula O2Si
UN Number NONH for all modes of transport
Packaging Metal Can
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.