Learn More
Miles Scientific SILICA GEL W/AGNO3 250UM 20X20CM (25 PLATES/BOX)
SDP
Catalog No. NC1398974
Encompass_Preferred
These silver nitrate impregnated silica gel plates are for argentation chromatography. They are impregnated with between 5% and 20% silver nitrate which modifies the separation characteristics of the adsorbent layer to permit increased discrimination of certain compounds, particularly those containing carbon-carbon double bonds. They are available with or without a fluorescent indicator. NOTE: The concentration of silver nitrate (5, 10, 15, or 20%) must be specified at the time of order. The most common concentration is 10%.
Catalog No. NC1398974 Supplier Miles Scientific Supplier No. P28011
These silver nitrate impregnated silica gel plates are for argentation chromatography. They are impregnated with between 5% and 20% silver nitrate which modifies the separation characteristics of the adsorbent layer to permit increased discrimination of certain compounds, particularly those containing carbon-carbon double bonds. They are available with or without a fluorescent indicator. NOTE: The concentration of silver nitrate (5, 10, 15, or 20%) must be specified at the time of order. The most common concentration is 10%.