Learn More
Miles Scientific SILICA GF 4000UM PRE-SCRAPED R
SDP
Catalog No. NC9468722
Encompass_Preferred
Rotor; Pre-scraped ready to use uniform layer; High performance silica gel GF 4000um; For CycloGraph and Chromatotron; UV254 Indicator; Precision cut glass
Catalog No. NC9468722 Supplier Miles Scientific Supplier No. R02206P
Rotor; Pre-scraped ready to use uniform layer; High performance silica gel GF 4000um; For CycloGraph and Chromatotron; UV254 Indicator; Precision cut glass