Learn More
Biochain Institute Inc PureSil-Silica Magnetic Beads, 10 mL/pk
SDP
Catalog No. 501871551
Encompass_Preferred
PureSil-Silica Magnetic Beads are Fe3O4 magnetic beads coated with a silicon dioxide (SiO2) layer. Since silica is able to bind to the nucleic acids, PureSil-Silica Magnetic Beads serve as a simple and efficient tool for plasmid DNA purification for transfection or sequencing applications, genomic DNA purification for research or clinical applications, RNA purification for qPCR analysis, or PCR product clean-up for downstream analysis.Features- Capacity: 4mg DNA/ml beads- 1-2 um particle size- Bead concentration: 4mg/ml- Bead density: ~1.03g/cm3- Simple, short procedure- No costly columns required- No time consuming centrifugations requiredApplications- Plasmid DNA isolation- Genomic DNA isolation- RNA purification for qPCR- PCR product clean up
Catalog No. 50-187-1551 Supplier Biochain Institute Inc Supplier No. L5011010
PureSil-Silica Magnetic Beads are Fe3O4 magnetic beads coated with a silicon dioxide (SiO2) layer. Since silica is able to bind to the nucleic acids, PureSil-Silica Magnetic Beads serve as a simple and efficient tool for plasmid DNA purification for transfection or sequencing applications, genomic DNA purification for research or clinical applications, RNA purification for qPCR analysis, or PCR product clean-up for downstream analysis.Features- Capacity: 4mg DNA/ml beads- 1-2 um particle size- Bead concentration: 4mg/ml- Bead density: ~1.03g/cm3- Simple, short procedure- No costly columns required- No time consuming centrifugations requiredApplications- Plasmid DNA isolation- Genomic DNA isolation- RNA purification for qPCR- PCR product clean up