Learn More
Cospheric LLC Monodisperse Silica Microspheres - 2.0g/cc D50= 9.20um, CV=4.8%, <1% Doubles - 1g
SDP
Catalog No. NC3790530
Encompass
Silica spheres are manufactured using proprietary technologies which allow the essential properties of amorphous silica to be maintained while simultaneously achieving controlled spherical shape and uniform particle size.Untreated silica spheres are typically hydrophilic and negatively charged. However, these parameters are not fixed, and might vary depending on the properties system the spheres are incorporated into.
Catalog No. NC3790530 Supplier Cospheric LLC Supplier No. SIO2MS2.0 9.2UM1G
Silica spheres are manufactured using proprietary technologies which allow the essential properties of amorphous silica to be maintained while simultaneously achieving controlled spherical shape and uniform particle size.Untreated silica spheres are typically hydrophilic and negatively charged. However, these parameters are not fixed, and might vary depending on the properties system the spheres are incorporated into.