Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silica Nanoparticles Non-porous 500 Nm 5% Wt/wt Ethanol Dispersion

Catalog No. 9136182ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-361-82ML 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-361-82ML Supplier Sigma Aldrich Supplier No. 9136182ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Monodisperse nonporous silica nanoparticles find application in drug delivery and molecular imaging whereas micron sized particles are used in liquid chromatography. We offer highly monodisperse nonporous silica in ethanol, with sizes varying from 150 nm to 900 nm. The high monodispersity of our particles makes them suitable for aforementioned applications.

Specifications

CAS 7631-86-9
Molecular Formula SiO2
Linear Formula SiO2
MDL Number MFCD00011232
Quantity 2 mL
Synonym Monodisperse silicon dioxide; Non-porous silica; Non-porous silicon dioxide; Silica
Molecular Weight (g/mol) 60.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.