Learn More
Sorbent Technologies Inc Silica NH2 HPTLC Plates, w/UV254, aluminum backed, 150um, 4x8cm, 50pk
SDP
Catalog No. NC1831712
Encompass_Preferred
This thin layer chromatography (TLC) plate comprises an Amino Silica layer. The neutral layer can be developed with purely organic, organic/aqueous and purely aqueous eluents. Thus, the phase finds numerous applications in normal phase chromatography as well as in the reversed phase mode. The Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 2-10um, and pH stability of 2-8. The TLC plates contain UV254 fluorescent indicator. This analytical layer is bound to an aluminum support. It has a thickness of 150um and dimensions of 4x8cm. There are 50 plates per pack. Applications include: Vitamins, sugars, steroids, purine derivatives, xanthines, phenols, nucleotides, and pesticides.
Catalog No. NC1831712 Supplier Sorbent Technologies Inc Supplier No. 3033187
This thin layer chromatography (TLC) plate comprises an Amino Silica layer. The neutral layer can be developed with purely organic, organic/aqueous and purely aqueous eluents. Thus, the phase finds numerous applications in normal phase chromatography as well as in the reversed phase mode. The Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 2-10um, and pH stability of 2-8. The TLC plates contain UV254 fluorescent indicator. This analytical layer is bound to an aluminum support. It has a thickness of 150um and dimensions of 4x8cm. There are 50 plates per pack. Applications include: Vitamins, sugars, steroids, purine derivatives, xanthines, phenols, nucleotides, and pesticides.