Learn More
Aqua Solutions Silica Reagent Set, Ultra Low Range Rapid Liquid
SDP
Catalog No. NC1915116
Encompass
For determination of ultra-low range Silica by the HeteropolCopper Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™y Blue Rapid Liquid™ method. Hach method 8282.Range: 3 - 1000 μg/L Set includes: Molybdate 3 Reagent Solution, Citric Acid F Reagent Solution, Amino Acid Reagent Dilution Solvent and Amino Acid F Reagent Powder for approximately 250 tests.Requires Pour-Thru Cell.(Customer mixes powder and diluent upon receipt)
Catalog No. NC1915116 Supplier Aqua Solutions Supplier No. HA2678500
For determination of ultra-low range Silica by the HeteropolCopper Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™y Blue Rapid Liquid™ method. Hach method 8282.Range: 3 - 1000 μg/L Set includes: Molybdate 3 Reagent Solution, Citric Acid F Reagent Solution, Amino Acid Reagent Dilution Solvent and Amino Acid F Reagent Powder for approximately 250 tests.Requires Pour-Thru Cell.(Customer mixes powder and diluent upon receipt)