Learn More
Nanocomposix NanoXact Gold Nanospheres - Silica Shelled - 60 nm, 1 mg/mL in ethanol, 1 mL
SDP
Supplier:  Nanocomposix AUSH201M
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.60 nm silica shelled spherical gold nanoparticles combine the optical properties of gold nanoparticles with the biocompatibility and functionalizability of a silica shell. These nanoparticles are useful in a broad range of applications from nanomedicine, targeted drug delivery, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. 50-195-9456