Learn More
Hach Company Silica Standard Solution, 10 mg/L as SiO2 (NIST), 500 mL
Catalog No. 501994554 Shop All Hach Company Products
Encompass
For accuracy checks in silica determinations. For use with Heteropoly Blue Method #8282.
Catalog No. 50-199-4554 Supplier Hach Company Supplier No. 140349
For accuracy checks in silica determinations. For use with Heteropoly Blue Method #8282.