Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Silica ULR Refill Kit
SDP

Catalog No. 13299707
Catalog No. 13-299-707 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R9011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Refill kit for Silica ULR visual test kits used for silica analysis in water. Contains 30 self-filling ampoules packaged securely in trays and a box.

  • Silica reacts with ammonium molybdate under acidic conditions to produce yellow molybdosilicic acid, which is reduced by aminonaphtholsulfonic acid to form heteropoly blue.
  • The resulting blue color is directly proportional to the silica concentration of the sample. Results are expressed as ppm (mg/L) SiO2. To convert test results to ppm Si, multiply by 0.467.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-9011
No. of Tests 30
Range 0 to 0.20 ppm
Type Silica ULR Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.