Learn More
CHEMetrics™ by AquaPhoenix Silica ULR Water Quality Visual Test Kit
SDP
Catalog No. 13299690
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-690 30
1 options
Catalog No. 13-299-690 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K9011
Add to Cart
Add to Cart
Silica ULR visual test kit supplied in a cardboard box. Contains everything needed to perform 30 tests.
  • Kit K-9011 comes in a cardboard box and contains everything needed to perform 30 tests: Refill, Comparator, Neutralizer Solution, Activator Solution, 25 mL sample cup, sample cup top, and instructions.
Specifications
Analytes Silica
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 0.20 ppm
Type ULR Water Quality Visual Test Kit