Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach™ Silica Ultra Low Range Bulk Reagent Set

Catalog No. 501996224 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Range:
3 to 1000μg/L SiO2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
50-199-6224 3 to 1000μg/L SiO2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-199-6224 Supplier Hach™ Supplier No. 2553500
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

For determination of ultra-low range Silica by the Heteropoly Blue Method. Hach™Silica Ultra Low Range Bulk Reagent Set includes: Molybdate 3, Citric Acid F Solution and Amino Acid F solution for approximately 100 tests.

TRUSTED_SUSTAINABILITY

Specifications

Analytes Silica (SiO2)
Format Bottle (Amino Acid F reagent) and Dropper Bottles
For Use With (Application) Ultra low range silica determination
For Use With (Equipment) Colorimeters (All Hach models except DR 900, DR/800 Series, PC II)
No. of Tests 100
Quantity 1 Set
Range 3 to 1000μg/L SiO2
Test Method Heteropoly blue
Type Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.