Learn More
Sorbent Technologies Inc Silica XG TLC Plates, w/UV254, aluminum backed, 200um, 2.5x7.5cm, 200pk
SDP
Catalog No. NC2881000
Encompass_Preferred
"Silica XG" is a standard grade Silica 60 with a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical soft layer is bound to an aluminum support. It has a thickness of 200um and dimensions of 2.5x7.5cm. There are 200 plates per pack. The Silica XG TLC Plate has outstanding wettability for precise colorization results, even with 100% aqueous detection reagents, and excellent separation efficiency and reproducibility from lot to lot. Cutting the plates is easy and reliable due to an optimized binder system. There is no flaking of silica gel layer. Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents.
Catalog No. NC2881000 Supplier Sorbent Technologies Inc Supplier No. 4434199
"Silica XG" is a standard grade Silica 60 with a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical soft layer is bound to an aluminum support. It has a thickness of 200um and dimensions of 2.5x7.5cm. There are 200 plates per pack. The Silica XG TLC Plate has outstanding wettability for precise colorization results, even with 100% aqueous detection reagents, and excellent separation efficiency and reproducibility from lot to lot. Cutting the plates is easy and reliable due to an optimized binder system. There is no flaking of silica gel layer. Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents.