Learn More
Medchemexpress LLC Silicic acid, magnesium salt | 1343-88-0 | 50 G
SDP
Catalog No. 5000411495
Encompass_Preferred
Magnesium silicate (activated magnesium silicate) is an orally active compound composed of magnesium oxide (MgO) and silicon dioxide (SiO2). It can be used in the preparation of food additives, antacids, and anti-ulcer agents. It also functions as a piezoelectric biomaterial, deodorant, decolorizing agent, and antifungal agent.
  • Releases magnesium and silicon ions
  • Promotes differentiation of mesenchymal stem cells into osteoblasts
  • Improves osteogenic properties when combined with various polymers
  • Neutralized in the stomach to form magnesium chloride and silicon dioxide
Catalog No. 50-004-11495 Supplier Medchemexpress LLC Supplier No. HYB220550G
Magnesium silicate (activated magnesium silicate) is an orally active compound composed of magnesium oxide (MgO) and silicon dioxide (SiO2). It can be used in the preparation of food additives, antacids, and anti-ulcer agents. It also functions as a piezoelectric biomaterial, deodorant, decolorizing agent, and antifungal agent.
  • Releases magnesium and silicon ions
  • Promotes differentiation of mesenchymal stem cells into osteoblasts
  • Improves osteogenic properties when combined with various polymers
  • Neutralized in the stomach to form magnesium chloride and silicon dioxide