Learn More
Allied High Tech Products SILICN CARBID DSC 600GRT 100/P
SDP
Catalog No. NC9937857
Encompass
8 in. (200mm) SILICON CARBIDE DISCS (Pk/100),Plain Back - 600 Grit,Features:Premium Resin Bonding System for longer wear rates,Durable C weight backing with latex additive for superior water resistance,Special adhesive backing
Catalog No. NC9937857 Supplier Allied High Tech Products Supplier No. 5010030
8 in. (200mm) SILICON CARBIDE DISCS (Pk/100),Plain Back - 600 Grit,Features:Premium Resin Bonding System for longer wear rates,Durable C weight backing with latex additive for superior water resistance,Special adhesive backing