Learn More
Med Vet International Silicon 6fr Gastric Feeding Tube, J0385D
Catalog No. 5000582048
Encompass_Preferred
Features: Bullet point/cap for luer fitting Lateral eye along numerous spots (4) Graduated marking Can be used long term Sterile
Catalog No. 50-005-82048 Supplier Med Vet International Supplier No. JOR385D
Features: Bullet point/cap for luer fitting Lateral eye along numerous spots (4) Graduated marking Can be used long term Sterile