Learn More
Zymo Research Corporation Silicon-A™ HRC Plate (2 Plates)
SDP
Catalog No. 501251326
Encompass_Preferred
Designed for efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from most impure DNA and RNA preparations. Must be prepared using Prep Solution before use.
Catalog No. 50-125-1326 Supplier Zymo Research Corporation Supplier No. C2009
Designed for efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from most impure DNA and RNA preparations. Must be prepared using Prep Solution before use.