Learn More
Zymo Research Corporation Silicon-A™ Plate (2 Plates)
SDP
Catalog No. 50444077
Encompass_Preferred
The Silicon-A Plate can be used in centrifuges for the large scale (i.e., 96-well) purification of DNA and/or RNA. Its lowprofile, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 g DNA or RNA in 30 l eluate per well. Capacity is 600 l per well.
Catalog No. 50-444-077 Supplier Zymo Research Corporation Supplier No. C2001
The Silicon-A Plate can be used in centrifuges for the large scale (i.e., 96-well) purification of DNA and/or RNA. Its lowprofile, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 g DNA or RNA in 30 l eluate per well. Capacity is 600 l per well.