Learn More
Allied High Tech Products Silicon Carbide Paper, 400 Grit, 8" (200 mm) Disc (Pk/100)
SDP
Supplier:  Allied High Tech Products 5010025
Encompass
Designed for metallographic applications to coarse and fine grind a wide variety of materials. They feature superior mineral grading, a unique resin top coat, and a latex additive in the paper. This combination resists water penetration, creating a durable, long-lasting disc with uniform cutting characteristics that minimizes sample distortion and deformation. Allied's silicon carbide abrasive discs are made with high quality C weight backing and are recommended for general laboratory requirements. Features: * Premium resin bonding system retains abrasive for sustained material removal and longer wear rates * Durable C weight backing with latex additive for superior water resistance
Catalog No. NC0290068