Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicon dioxide

Catalog No. 502907993
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7993 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-7993 Supplier Sigma Aldrich Supplier No. 637238500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicon dioxide
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2230°C
Melting Point >1600°C
CAS 7631-86-9
Quantity 500 g
Molecular Formula O2Si
Linear Formula SiO2
MDL Number MFCD00011232
Synonym Silica; Silicon dioxide
Molecular Weight (g/mol) 60.08
Percent Purity 99.5% trace metals basis
Density 2.2-2.6 g/mL (at 25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.