Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Dioxide, calcined, crude, Solstice

Catalog No. 60045064 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
50 kg
6 x 2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-064 6 x 2.5 kg
60-020-38 2.5 kg
60-020-39 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-064 Supplier Solstice Supplier No. 186486X2.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for cleaning of platinum crucibles, calcined, crude

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 6 x 2.5 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08
Chemical Name or Material Silicon Dioxide
EINECSNumber 238-878-4
RTECSNumber VV7330000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.