Learn More
Abcam Silicon Imaging Chamber (1.75 mm), 6UNIT
Catalog No. 502672468
Encompass_Preferred
After you have finished staining and clearing your tissues using Abcam's Tissue Clearing Kits and reagents, you can use Silicon Imaging Chambers to mount your tissue for imaging.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2468 Supplier Abcam Supplier No. AB2433096UNIT
After you have finished staining and clearing your tissues using Abcam's Tissue Clearing Kits and reagents, you can use Silicon Imaging Chambers to mount your tissue for imaging.
The product is subject to the following: Abcam Restricted Use Statement