Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) oxide sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)

Catalog No. AA41099KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41099KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41099KS Supplier Thermo Scientific Chemicals Supplier No. 041099KS
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

Silicon(IV) oxide sputtering target has low thermal expansion coefficient which makes it ideal for mirrors and optical flats. It is used for viewing windows, being transparent to wavelengths from around 0.2 to 3.5 microns, insulators for electronic applications and for semi-conductor manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011232

Specifications

Quantity 1 Ea.
Solubility Information Insoluble in water.
Formula Weight 60.09
Percent Purity 99.995%
Assay (metals basis)
Chemical Name or Material Silicon(IV) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.