Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Metal, Lumps
SCI_ED

Catalog No. S93348 Shop All Fisherbrand Products
Change view
Click to view available options
Metal; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S93348 Metal; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S93348

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Silver to Gray
CAS 7440-21-3
Quantity 500 g
Molecular Formula Si
Linear Formula Si
MDL Number MFCD00085311
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES [Si]
IUPAC Name silicon
Molecular Weight (g/mol) 28.09
PubChem CID 5461123
ChEBI CHEBI:27573
Formula Weight 28.09
Packaging Metal
Chemical Name or Material Silicon Metal, Lumps
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife Indefinite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.