Learn More
Electron Microscopy Sciences EMS Silicon Monoxide Film on on 300 mesh Nickel Grid
SDP
Catalog No. 5033099
Encompass
A thin film of pure silicone Monoxide (15 – 30 nm) is deposited directly on top of the grid. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Catalog No. 50-330-99 Supplier Electron Microscopy Sciences Supplier No. SF300NI
A thin film of pure silicone Monoxide (15 – 30 nm) is deposited directly on top of the grid. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel