Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicon nitride

Catalog No. 334103100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-410-3100G 100 g
33-410-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-410-3100G Supplier Sigma Aldrich Supplier No. 334103100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicon nitride based ceramics possess high fracture toughness, hardness and wear resistance, resistance to corrosion, thermal stability. Two known polymorphs of silicon nitride exist, alpha and beta Si3N4, both have hexagonal structure. A study shows that a third state, cubic spinel structure, has also been synthesized.

Specifications

CAS 12033-89-5
Quantity 100 g
Molecular Formula N4Si3
Linear Formula Si3N4
MDL Number MFCD00011230
Synonym Trisilicon tetranitride
Molecular Weight (g/mol) 140.28
Percent Purity ≥99.9% trace metals basis
Density 3.44 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.