Learn More
Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL
Description
Silicon plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Solubility | Miscible with molten alkali oxides, a mixture of nitric and hydrofluoric acids; Immiscible with water, nitric and hydrochloric acid |
| Type | Analytical Standard |
| Quantity | 500 mL |
| Grade | Specpure |
| Chemical Name or Material | Silicon plasma standard solution, Si 10000μg/mL |
Safety and Handling
| Hazard Category | H302 |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.