Learn More
Acs Material SILICON/SILICON DIOXIDE WAFER
SDP
Catalog No. NC1875706
Encompass
CAS No.: 7440-21-3 / 7631-86-9 Si/SiO2 wafers from ACS Material are the industry standard for high-quality substrates. Our high-quality violet wafers are packaged in a 1000 class cleanroom and provide optimal visibility for a variety of nanomaterials, including CVD graphene and graphene flakes.
Catalog No. NC1875706 Supplier Acs Material Supplier No. WSOP3004
CAS No.: 7440-21-3 / 7631-86-9 Si/SiO2 wafers from ACS Material are the industry standard for high-quality substrates. Our high-quality violet wafers are packaged in a 1000 class cleanroom and provide optimal visibility for a variety of nanomaterials, including CVD graphene and graphene flakes.