Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Standard, 1mL = 1mg Si, 1000ppm Si, Ricca Chemical
SDP

Catalog No. 68004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
68004 120 mL Poly natural
6800-16 500 mL Poly natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68004 Supplier Ricca Chemical Company Supplier No. 68004

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6834-92-0 , 7732-18-5
Molecular Formula Na2O3Si
Molecular Weight (g/mol) 122.062
InChI Key NTHWMYGWWRZVTN-UHFFFAOYSA-N
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Si (2139ppm SiO2)
Equivalence 1 mL = 1 mg Si
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Silicon Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.