Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Reinforced Peroxide-Cured Silicone, 1/4"" ID x 9/16"" OD; 25 ft
Catalog No. NC1290620
Encompass
Applications: Demanding high-pressure pharmaceutical, biotechnology, aerospace, and food and beverage applications. Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, low tackiness, translucent. Certifications: Elastomer is FDA compliant and meets USP Class VI. Tubing is manufactured according to GMP. Temperature range: -60 to 400¿F (-51 to 204¿C) Sterilize: Autoclave or gamma irradiation.
Catalog No. NC1290620 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9562303
Applications: Demanding high-pressure pharmaceutical, biotechnology, aerospace, and food and beverage applications. Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, low tackiness, translucent. Certifications: Elastomer is FDA compliant and meets USP Class VI. Tubing is manufactured according to GMP. Temperature range: -60 to 400¿F (-51 to 204¿C) Sterilize: Autoclave or gamma irradiation.