Learn More
Electron Microscopy Sciences Silicon Test Specimen, Incident L.M.
SDP
Catalog No. 5028609
Encompass_Preferred
This Test Specimen is made of a 5 mmx5 mm square of single crystal silicon. It is photo-etched and the squares repeat every 10 um. The dividing lines are 1.9 um wide.
A broader etching line is written every 500 um, which is useful in light microscopy. This is an excellent test specimen for comparing magnification and assessing any distortion in the image field. It is ideally useful in the context of automated counting systems to check for unsuspected distortions. Where critical measurements must be made the sample can be mounted directly onto the calibration specimen so that an internal calibration is obtained on the micro-graph.
Catalog No. 50-286-09 Supplier Electron Microscopy Sciences Supplier No. 7950220
This Test Specimen is made of a 5 mmx5 mm square of single crystal silicon. It is photo-etched and the squares repeat every 10 um. The dividing lines are 1.9 um wide.
A broader etching line is written every 500 um, which is useful in light microscopy. This is an excellent test specimen for comparing magnification and assessing any distortion in the image field. It is ideally useful in the context of automated counting systems to check for unsuspected distortions. Where critical measurements must be made the sample can be mounted directly onto the calibration specimen so that an internal calibration is obtained on the micro-graph.