Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicon tetrachloride

Catalog No. 289388100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-938-8100ML 100 mL
28-938-8800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-938-8100ML Supplier Sigma Aldrich Supplier No. 289388100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicon tetrachloride

Specifications

CAS 10026-04-7
Melting Point -70°C
Density 1.483 g/mL (at 25°C)
Boiling Point 57.6°C
Molecular Formula Cl4Si
Linear Formula SiCl4
Quantity 100 mL
MDL Number MFCD00011229
Synonym STC; Tetrachlorosilane
Molecular Weight (g/mol) 169.9
Percent Purity 99.998% trace metals basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.