Learn More
Helix Medical Inc SILICON TUBING 50 FT .025X.047
Catalog No. NC0518633
Encompass_Preferred
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations.
Catalog No. NC0518633 Supplier Helix Medical Inc Supplier No. 6001103
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations.