Learn More
Addison Engineering Inc SILICON WAFER 4IN 4P009/24CS
SDP
Catalog No. NC0227985
Encompass_Preferred
4 in. diameter prime grade silicon wafers, single side polished, case of 24 wafers
Catalog No. NC0227985 Supplier Addison Engineering Inc Supplier No. 4P009CS
4 in. diameter prime grade silicon wafers, single side polished, case of 24 wafers