Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, 1/8 in ID, 1/4 in OD
Catalog No. NC3312429
Encompass_Preferred
Platinum-cured, ultra-pure silicone tubing with ultra-low extractables. 0.125 in (3.18 mm) inner diameter and 0.25 in (6.35 mm) outer diameter supplied as a 50 ft coil with 50 Shore A hardness. Designed for biopharmaceutical and laboratory fluid transfer, it offers extended pump life in peristaltic applications.
  • Platinum-cured silicone with ultra-low extractables.
  • Inner diameter 0.13 in (3.18 mm), outer diameter 0.25 in (6.35 mm), wall thickness 1/16 in (1.59 mm).
  • Length 50 ft coil.
  • Hardness 50 Shore A.
  • Temperature range -80°F (-62°C) to 500°F (260°C).
  • Engineered for peristaltic pumps and single-use bioprocess assemblies.
Catalog No. NC3312429 Supplier Accuflow Systems Supplier No. ULTRAC1252F
Platinum-cured, ultra-pure silicone tubing with ultra-low extractables. 0.125 in (3.18 mm) inner diameter and 0.25 in (6.35 mm) outer diameter supplied as a 50 ft coil with 50 Shore A hardness. Designed for biopharmaceutical and laboratory fluid transfer, it offers extended pump life in peristaltic applications.
  • Platinum-cured silicone with ultra-low extractables.
  • Inner diameter 0.13 in (3.18 mm), outer diameter 0.25 in (6.35 mm), wall thickness 1/16 in (1.59 mm).
  • Length 50 ft coil.
  • Hardness 50 Shore A.
  • Temperature range -80°F (-62°C) to 500°F (260°C).
  • Engineered for peristaltic pumps and single-use bioprocess assemblies.