Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Platinum-Cured, 0.25 in ID, 50 ft
Catalog No. NC3312432
Encompass_Preferred
Ultra-pure platinum-cured silicone tubing for biopharmaceutical and laboratory fluid transfer, supplied as a 50 ft coil with 0.25 in (6.35 mm) inner diameter and 1/16 in wall thickness.
  • Platinum-cured silicone offering low extractables and long pump life.
  • 0.25 in (6.35 mm) inner diameter, 1/16 in wall thickness (≈0.375 in outer diameter).
  • 50 Shore A hardness balances flexibility and pump durability.
  • Wide temperature range: -80°F (-62°C) to 500°F (260°C).
  • Manufactured and packaged in a Class 7 clean room to minimize contamination.
  • Suitable for peristaltic pumps, single-use assemblies, and fluid transfer in pharmaceutical and bioprocessing applications.
Catalog No. NC3312432 Supplier Accuflow Systems Supplier No. ULTRAC2502F
Ultra-pure platinum-cured silicone tubing for biopharmaceutical and laboratory fluid transfer, supplied as a 50 ft coil with 0.25 in (6.35 mm) inner diameter and 1/16 in wall thickness.
  • Platinum-cured silicone offering low extractables and long pump life.
  • 0.25 in (6.35 mm) inner diameter, 1/16 in wall thickness (≈0.375 in outer diameter).
  • 50 Shore A hardness balances flexibility and pump durability.
  • Wide temperature range: -80°F (-62°C) to 500°F (260°C).
  • Manufactured and packaged in a Class 7 clean room to minimize contamination.
  • Suitable for peristaltic pumps, single-use assemblies, and fluid transfer in pharmaceutical and bioprocessing applications.