Learn More
Accuflow Systems Saint-Gobain Sani-Tech Ultra-C Tubing, 0.375 in ID, 0.75 in OD
Catalog No. NC3312436
Encompass_Preferred
Sani-Tech Ultra-C platinum-cured silicone tubing provides ultra-low extractables and extended pump life for critical biopharmaceutical fluid-transfer applications. It is configured with a 0.375 in (9.53 mm) inner diameter, 0.75 in (19.05 mm) outer diameter, and supplied on a 50 ft spool.
  • Platinum-cured silicone construction with ultra-low extractables.
  • 0.375 in (9.53 mm) inner diameter, 0.75 in (19.05 mm) outer diameter.
  • 50 ft (15.24 m) length spool.
  • 50 Shore A hardness for flexible yet durable performance.
  • Wide temperature range: -80°F (-62°C) to 500°F (260°C) for sterilization and cryogenic use.
  • Suitable for biopharmaceutical and critical fluid contact applications requiring high fluid integrity.
Catalog No. NC3312436 Supplier Accuflow Systems Supplier No. ULTRAC3755F
Sani-Tech Ultra-C platinum-cured silicone tubing provides ultra-low extractables and extended pump life for critical biopharmaceutical fluid-transfer applications. It is configured with a 0.375 in (9.53 mm) inner diameter, 0.75 in (19.05 mm) outer diameter, and supplied on a 50 ft spool.
  • Platinum-cured silicone construction with ultra-low extractables.
  • 0.375 in (9.53 mm) inner diameter, 0.75 in (19.05 mm) outer diameter.
  • 50 ft (15.24 m) length spool.
  • 50 Shore A hardness for flexible yet durable performance.
  • Wide temperature range: -80°F (-62°C) to 500°F (260°C) for sterilization and cryogenic use.
  • Suitable for biopharmaceutical and critical fluid contact applications requiring high fluid integrity.