Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Ultra-Pure Platinum-Cured, 0.38 in ID, 50 ft
Catalog No. NC3312434
Encompass_Preferred
Saint-Gobain Sani-Tech Ultra-C silicone tubing is an ultra-pure, platinum-cured silicone tube engineered for biopharmaceutical and laboratory fluid transfer applications. It provides low extractables, extended pump life, and compatibility with repeated autoclaving for sterile processing, media handling, and fill/finish operations.
  • Ultra-pure platinum-cured silicone construction.
  • 0.38 in (9.53 mm) inner diameter.
  • 0.63 in (15.88 mm) outer diameter.
  • 0.13 in (3.18 mm) wall thickness.
  • 50 ft (15.24 m) coil length.
  • 50 Shore A hardness.
  • Temperature range -80 °F (-62 °C) to 500 °F (260 °C).
  • Meets USP <88> Class VI, USP <87>, ISO 10993-5, and European Pharmacopoeia 3.1.9.
  • Typical applications: sterile fill and transfers, media/buffer processing, pumping, cell culture, chromatography, filtration, and fermentation.
Catalog No. NC3312434 Supplier Accuflow Systems Supplier No. ULTRAC3754F
Saint-Gobain Sani-Tech Ultra-C silicone tubing is an ultra-pure, platinum-cured silicone tube engineered for biopharmaceutical and laboratory fluid transfer applications. It provides low extractables, extended pump life, and compatibility with repeated autoclaving for sterile processing, media handling, and fill/finish operations.
  • Ultra-pure platinum-cured silicone construction.
  • 0.38 in (9.53 mm) inner diameter.
  • 0.63 in (15.88 mm) outer diameter.
  • 0.13 in (3.18 mm) wall thickness.
  • 50 ft (15.24 m) coil length.
  • 50 Shore A hardness.
  • Temperature range -80 °F (-62 °C) to 500 °F (260 °C).
  • Meets USP <88> Class VI, USP <87>, ISO 10993-5, and European Pharmacopoeia 3.1.9.
  • Typical applications: sterile fill and transfers, media/buffer processing, pumping, cell culture, chromatography, filtration, and fermentation.