Learn More
Med Vet International Silicone 7.0mm Endotracheal Tube with Wire Reinforcement, J0629F
Catalog No. 5000581474
Encompass_Preferred
The tube has a metal wire reinforcement that spirals the length of the tube. This reinforcement gives rigidity and prevents any possible occlusion. Features: Graduations to aid in depth placement. Radiopaque line for location confirmation. Fully transparent to allow observation for development of blood or mucus. Lateral Murphy eye.
Catalog No. 50-005-81474 Supplier Med Vet International Supplier No. JOR629F
The tube has a metal wire reinforcement that spirals the length of the tube. This reinforcement gives rigidity and prevents any possible occlusion. Features: Graduations to aid in depth placement. Radiopaque line for location confirmation. Fully transparent to allow observation for development of blood or mucus. Lateral Murphy eye.