Learn More
Crescent Chemical Co Inc SILICONE ANTI-FOAM EMULSION
SDP
Catalog No. 50248136
Encompass_Preferred
Silicone anti-foam emulsion, 30 % USP Dow Corning 30 % polydimethylsiloxane in water; contains traces of emulsifiers derived from plant. Size - 100ML Storage Conditions - +15 C TO +30 C Catalog Number - 35119.01
Catalog No. 50-248-136 Supplier Crescent Chemical Co Inc Supplier No. SE35119.01
Silicone anti-foam emulsion, 30 % USP Dow Corning 30 % polydimethylsiloxane in water; contains traces of emulsifiers derived from plant. Size - 100ML Storage Conditions - +15 C TO +30 C Catalog Number - 35119.01