Learn More
Electron Microscopy Sciences mPrep™ Bench, 96-well rack, silicone
SDP
Catalog No. 501928124
Encompass_Preferred
mPrep System Accessory compatible with mPrep/s and mPrep/g. 96-well silicone rack provides tight seal to capsule bottoms during incubations or transfer of fluid-filled capsules to ovens and incubators. Autoclavable. Compatible with mPrep/s and mPrep/g.
Catalog No. 50-192-8124 Supplier Electron Microscopy Sciences Supplier No. 8501016
mPrep System Accessory compatible with mPrep/s and mPrep/g. 96-well silicone rack provides tight seal to capsule bottoms during incubations or transfer of fluid-filled capsules to ovens and incubators. Autoclavable. Compatible with mPrep/s and mPrep/g.