Learn More
Analytical Sales & Services Silicone Cap Mat for 8x30mm Vials in 24 Well Plate (9mm Spacing) Format
Catalog No. 5000806913
Encompass
Advantage Silicone Cap Mat for 1mL, 8x30mm Vials in 24 Well Format\n\n• 9mm spacing\n\nNote: this is a 96-well format cap mat (99685) cut into 4 equal quadrants \n\nQty/Pkg: 20 (24-well sections)
Catalog No. 50-008-06913 Supplier Analytical Sales & Services Supplier No. 996854
Advantage Silicone Cap Mat for 1mL, 8x30mm Vials in 24 Well Format\n\n• 9mm spacing\n\nNote: this is a 96-well format cap mat (99685) cut into 4 equal quadrants \n\nQty/Pkg: 20 (24-well sections)