Learn More
Analytical Sales & Services Silicone Cap Mat for 96 Position Radiochemistry Reactor
Catalog No. 5000806464
Encompass
Silicone Cap Mat for 96 Position Radiochemistry Reactor\n\n• For 8x30mm glass insert vials in Radiochemistry Reactor (96950) or  Conversion Kit (96940)\n• Seals the vials during experimentation\n\nQty/Pkg: 5
Catalog No. 50-008-06464 Supplier Analytical Sales & Services Supplier No. 96958
Add to Cart
Add to Cart
Silicone Cap Mat for 96 Position Radiochemistry Reactor\n\n• For 8x30mm glass insert vials in Radiochemistry Reactor (96950) or  Conversion Kit (96940)\n• Seals the vials during experimentation\n\nQty/Pkg: 5