Learn More
Electron Microscopy Sciences Wet or Dry Silicone Carbide Disc 8", 320 Grit, Plain back
SDP
Catalog No. 50997147
Encompass_Preferred
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 320 Grit
Backing: Plain
Catalog No. 50-997-147 Supplier Electron Microscopy Sciences Supplier No. 5025006
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 320 Grit
Backing: Plain