Learn More
Electron Microscopy Sciences Wet or Dry Silicone Carbide Disc 8", 1200 Grit, PSA backing
SDP
Catalog No. 50997160
Encompass_Preferred
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 1200 Grit
Backing: PSA
Catalog No. 50-997-160 Supplier Electron Microscopy Sciences Supplier No. 5025510
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 1200 Grit
Backing: PSA